Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DDX3X SIRNA Assays ›
          Gene Symbol:
          Ddx3x
          Gene Name:
          DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 3, X-linked
          Gene Aliases:
          D1Pas1-rs2, Ddx3, Fin14
          Chromosome Location:
          Chr.X: 13280496 - 13293983 on Build GRCm38
          UniGene ID:
          Mm.289662
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s64904

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 368.00
          Your Price
          Online offer:
          368.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ddx3x
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010028.3 NP_034158.1 14 1653
          GenBank mRNA AK140990.1 - 14 1654
          AK163250.1 - 14 1628
          BC150862.1 - 14 1607 9087773
          BC172016.1 - 14 1604 9054302
          L25126.1 - 14 1585
          Z38117.1 - 14 1637

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001108246, XM_006256652, XM_006256653, FQ211849, BC085914.

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          translational initiation
          regulation of translation
          apoptotic process
          chromosome segregation
          extrinsic apoptotic signaling pathway via death domain receptors
          response to virus
          regulation of gene expression
          RNA secondary structure unwinding
          positive regulation of gene expression
          negative regulation of translation
          positive regulation of cell growth
          negative regulation of cell growth
          negative regulation of protein complex assembly
          DNA duplex unwinding
          stress granule assembly
          intracellular signal transduction
          ribosome biogenesis
          mature ribosome assembly
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of viral genome replication
          innate immune response
          positive regulation of translation
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of translational initiation
          cellular response to arsenic-containing substance
          cellular response to osmotic stress
          intrinsic apoptotic signaling pathway
          positive regulation of G1/S transition of mitotic cell cycle
          negative regulation of intrinsic apoptotic signaling pathway
          nucleotide binding
          nucleic acid binding
          DNA binding
          RNA binding
          GTPase activity
          ATP-dependent DNA helicase activity
          ATP-dependent RNA helicase activity
          helicase activity
          protein binding
          ATP binding
          transcription factor binding
          poly(A) binding
          eukaryotic initiation factor 4E binding
          hydrolase activity
          ATPase activity
          nucleoside-triphosphatase activity
          translation initiation factor binding
          RNA stem-loop binding
          ribosomal small subunit binding
          CTPase activity
          poly(A) RNA binding
          mRNA 5'-UTR binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0