Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CAV3 SIRNA Assays ›
          Gene Symbol:
          Cav3
          Gene Name:
          caveolin 3
          Gene Aliases:
          AI385751, Cav-3, M-cav
          Chromosome Location:
          Chr.6: 112459505 - 112472872 on Build GRCm38
          UniGene ID:
          Mm.3924
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s63431

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Cav3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007617.3 NP_031643.1 2 473
          GenBank mRNA BC024383.1 - 2 483 4988598
          CJ057698.1 - 2 250
          U36579.1 - 2 287

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          plasma membrane repair
          regulation of heart rate
          negative regulation of protein kinase activity
          triglyceride metabolic process
          endocytosis
          plasma membrane organization
          actin filament organization
          positive regulation of cytosolic calcium ion concentration
          myoblast fusion
          regulation of heart contraction
          protein localization
          positive regulation of cell proliferation
          negative regulation of cardiac muscle hypertrophy
          positive regulation of myotube differentiation
          regulation of skeletal muscle contraction
          myotube differentiation
          regulation of transforming growth factor beta receptor signaling pathway
          positive regulation of microtubule polymerization
          cytoplasmic microtubule organization
          membrane raft organization
          detection of muscle stretch
          regulation of signal transduction by receptor internalization
          regulation of membrane potential
          glucose homeostasis
          cholesterol homeostasis
          negative regulation of MAP kinase activity
          negative regulation of MAPK cascade
          negative regulation of cell size
          muscle cell cellular homeostasis
          negative regulation of nitric-oxide synthase activity
          regulation of nerve growth factor receptor activity
          nucleus localization
          regulation of protein kinase B signaling
          regulation of calcium ion transport
          negative regulation of calcium ion transport
          cardiac muscle cell development
          regulation of cardiac muscle contraction
          negative regulation of sarcomere organization
          regulation of ventricular cardiac muscle cell membrane repolarization
          heart trabecula formation
          regulation of ventricular cardiac muscle cell membrane depolarization
          regulation of branching involved in mammary gland duct morphogenesis
          negative regulation of cell growth involved in cardiac muscle cell development
          caveola assembly
          protein localization to plasma membrane
          ventricular cardiac muscle cell action potential
          establishment of protein localization to plasma membrane
          regulation of calcium ion import
          regulation of p38MAPK cascade
          regulation of membrane depolarization during cardiac muscle cell action potential
          negative regulation of membrane depolarization during cardiac muscle cell action potential
          negative regulation of potassium ion transmembrane transporter activity
          regulation of calcium ion transmembrane transporter activity
          negative regulation of potassium ion transmembrane transport
          regulation of sodium ion transmembrane transporter activity
          positive regulation of caveolin-mediated endocytosis
          calcium channel regulator activity
          protein binding
          protein C-terminus binding
          sodium channel regulator activity
          potassium channel inhibitor activity
          enzyme binding
          protein complex binding
          protein complex scaffold
          alpha-tubulin binding
          ion channel binding
          nitric-oxide synthase binding
          connexin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-956b84784-lf4xt:80/100.66.134.75:80.
          git-commit: 47437712f32dbf53ebfc670c484059c2aca19dcb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0