Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CEP170 SIRNA Assays ›
          Gene Symbol:
          CEP170
          Gene Name:
          centrosomal protein 170
          Gene Aliases:
          FAM68A, KAB, KIAA0470
          Chromosome Location:
          Chr.1: 243124428 - 243255936 on Build GRCh38
          UniGene ID:
          Hs.533635
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s54936

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CEP170
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042404.1 NP_001035863.1 12 3362
          NM_001042405.1 NP_001035864.1 12 3362
          NM_014812.2 NP_055627.2 13 3656
          XM_006711843.3 XP_006711906.1 12 4365
          XM_011544334.2 XP_011542636.1 12 4372
          XM_011544335.2 XP_011542637.1 12 3785
          XM_011544336.2 XP_011542638.1 12 4691
          XM_011544337.2 XP_011542639.1 12 4372
          XM_011544338.2 XP_011542640.1 12 4372
          XM_011544339.2 XP_011542641.1 12 4372
          XM_011544340.2 XP_011542642.1 12 4322
          XM_011544341.2 XP_011542643.1 13 4136
          XM_011544342.2 XP_011542644.1 12 4322
          XM_011544343.2 XP_011542645.1 11 3845
          XM_011544344.2 XP_011542646.1 12 3892
          XM_017002932.1 XP_016858421.1 12 4285
          XM_017002933.1 XP_016858422.1 13 3956
          XM_017002934.1 XP_016858423.1 13 3599
          XM_017002935.1 XP_016858424.1 13 4136
          XM_017002936.1 XP_016858425.1 13 3599
          XM_017002937.1 XP_016858426.1 13 4136
          XM_017002938.1 XP_016858427.1 13 4136
          XM_017002939.1 XP_016858428.1 13 3956
          XM_017002940.1 XP_016858429.1 13 3599
          XM_017002941.1 XP_016858430.1 13 3956
          XM_017002942.1 XP_016858431.1 13 4136
          XM_017002943.1 XP_016858432.1 13 3956
          XM_017002944.1 XP_016858433.1 12 3885
          XM_017002945.1 XP_016858434.1 12 3892
          XM_017002946.1 XP_016858435.1 12 3659
          XM_017002947.1 XP_016858436.1 12 3892
          XM_017002948.1 XP_016858437.1 12 3885
          XM_017002949.1 XP_016858438.1 12 3892
          XM_017002950.1 XP_016858439.1 12 3659
          XM_017002951.1 XP_016858440.1 11 3365
          GenBank mRNA AB007939.1 - 11 3007
          AB022657.1 - 12 3301
          AB022658.1 - 11 3007
          AB022659.1 - 11 3007
          AK001664.1 - 1 944
          BC140794.1 - 11 2998 9021662
          BC146790.1 - 11 3007 9007254

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0