Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ATP13A2 SIRNA Assays ›
          Gene Symbol:
          ATP13A2
          Gene Name:
          ATPase 13A2
          Gene Aliases:
          CLN12, HSA9947, KRPPD, PARK9
          Chromosome Location:
          Chr.1: 16985958 - 17011972 on Build GRCh38
          UniGene ID:
          Hs.128866
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s536648

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ATP13A2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001141973.2 NP_001135445.1 14, 15 1579
          NM_001141974.2 NP_001135446.1 14, 15 1579
          NM_022089.3 NP_071372.1 14, 15 1594
          XM_005245810.1 XP_005245867.1 14, 15 1591
          XM_005245811.1 XP_005245868.1 14, 15 1579
          XM_005245812.1 XP_005245869.1 14, 15 1567
          XM_005245815.1 XP_005245872.1 14, 15 1594
          XM_006710512.1 XP_006710575.1 14, 15 1576
          XM_006710513.1 XP_006710576.1 14, 15 1552
          XM_011541128.1 XP_011539430.1 14, 15 1594
          XM_011541129.1 XP_011539431.1 14, 15 1594
          XM_017000844.1 XP_016856333.1 14, 15 1594
          XM_017000845.1 XP_016856334.1 14, 15 1576
          XM_017000846.1 XP_016856335.1 14, 15 1552
          XM_017000847.1 XP_016856336.1 14, 15 1564
          XM_017000848.1 XP_016856337.1 14, 15 1594
          XM_017000849.1 XP_016856338.1 14, 15 1579
          XM_017000850.1 XP_016856339.1 14, 15 1594
          GenBank mRNA AK075310.1 - 4, 5 376
          AK290210.1 - 14, 15 1535
          AL354615.1 - 14, 15 1394
          AY461712.1 - 14, 15 1393
          AY987009.1 - 4, 5 388
          BC034575.1 - 11, 12 1085

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cellular calcium ion homeostasis
          cellular iron ion homeostasis
          cellular zinc ion homeostasis
          regulation of mitochondrion organization
          regulation of macroautophagy
          regulation of autophagosome size
          cellular cation homeostasis
          regulation of intracellular protein transport
          ion transmembrane transport
          cellular response to oxidative stress
          protein autophosphorylation
          positive regulation of protein secretion
          regulation of endopeptidase activity
          zinc ion homeostasis
          cellular response to manganese ion
          cellular response to zinc ion
          cation transmembrane transport
          negative regulation of neuron death
          regulation of mitophagy
          positive regulation of exosomal secretion
          regulation of chaperone-mediated autophagy
          autophagosome organization
          polyamine import
          regulation of lysosomal protein catabolic process
          negative regulation of lysosomal protein catabolic process
          peptidyl-aspartic acid autophosphorylation
          protein binding
          ATP binding
          zinc ion binding
          ATPase activity
          cation-transporting ATPase activity
          manganese ion binding
          phosphatidic acid binding
          phosphatidylinositol-3,5-bisphosphate binding
          cupric ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-858577f6d7-cpd7z:80/100.66.134.75:80.
          git-commit: 8a4c7f5d60a2609f21a9257632c46d3db2b553cc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0