Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other COBL SIRNA Assays ›
          Gene Symbol:
          COBL
          Gene Name:
          cordon-bleu WH2 repeat protein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 51016212 - 51316818 on Build GRCh38
          UniGene ID:
          Hs.99141
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s53096

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: COBL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001287436.1 NP_001274365.1 3 570
          NM_001287438.1 NP_001274367.1 3 570
          NM_015198.3 NP_056013.2 3 570
          XM_005271750.1 XP_005271807.1 3 561
          XM_005271751.1 XP_005271808.1 3 561
          XM_005271756.1 XP_005271813.1 3 561
          XM_011515234.1 XP_011513536.1 3 561
          XM_011515235.1 XP_011513537.1 3 561
          XM_011515236.1 XP_011513538.1 3 561
          XM_011515237.1 XP_011513539.1 3 561
          XM_011515238.1 XP_011513540.1 3 561
          XM_011515239.1 XP_011513541.1 3 561
          XM_011515240.1 XP_011513542.1 3 561
          XM_017011898.1 XP_016867387.1 3 561
          XM_017011899.1 XP_016867388.1 3 561
          XM_017011900.1 XP_016867389.1 3 561
          XM_017011901.1 XP_016867390.1 3 561
          GenBank mRNA AB014533.1 - 3 551
          AK309254.1 - 3 561
          BC094695.1 - 3 516
          BC111496.1 - 3 570
          BC136441.1 - 3 485 9020430
          BC144099.1 - 3 485 9052619
          BC150263.1 - 3 551 9007207
          DA245843.1 - 3 437

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          embryonic axis specification
          somite specification
          neural tube closure
          liver development
          actin filament polymerization
          notochord development
          floor plate development
          digestive tract development
          collateral sprouting in absence of injury
          actin filament network formation
          positive regulation of dendrite development
          positive regulation of ruffle assembly
          actin monomer binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0