Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIGA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270384.1 | NP_001257313.1 | 16 | 1768 | ||
NM_198549.3 | NP_940951.1 | 16 | 1765 | |||
XM_005270846.4 | XP_005270903.1 | 15 | 1675 | |||
XM_005270847.4 | XP_005270904.1 | 16 | 2446 | |||
XM_005270848.4 | XP_005270905.1 | 16 | 2055 | |||
XM_011541422.2 | XP_011539724.1 | 16 | 1819 | |||
XM_017001227.1 | XP_016856716.1 | 16 | 1735 | |||
XM_017001228.1 | XP_016856717.1 | 16 | 2052 | |||
XM_017001229.1 | XP_016856718.1 | 16 | 1827 | |||
XM_017001230.1 | XP_016856719.1 | 16 | 1840 | |||
GenBank mRNA | AK092412.1 | - | 16 | 1733 | ||
AK296411.1 | - | 15 | 1629 | |||
BC130387.1 | - | 16 | 1753 | 40146418 | ||
BC144167.1 | - | 16 | 1756 | 9052687 | ||
BX537792.1 | - | 16 | 1765 |