Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMEM218 SIRNA Assays ›
          Gene Symbol:
          TMEM218
          Gene Name:
          transmembrane protein 218
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 125094370 - 125111763 on Build GRCh38
          UniGene ID:
          Hs.731899
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s47734

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TMEM218
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001080546.2 NP_001074015.1 4, 5 659
          NM_001258238.1 NP_001245167.1 4, 5 537
          NM_001258239.1 NP_001245168.1 4, 5 634
          NM_001258240.1 NP_001245169.1 6, 7 697
          NM_001258241.1 NP_001245170.1 5, 6 638
          NM_001258242.1 NP_001245171.1 4, 5 625
          NM_001258243.1 NP_001245172.1 5, 6 728
          NM_001258244.1 NP_001245173.1 4, 5 528
          NM_001258245.1 NP_001245174.1 6, 7 690
          NM_001258246.1 NP_001245175.1 4, 5 610
          NM_001258247.1 NP_001245176.1 4, 5 513
          NR_047586.1 4, 5 490
          NR_047587.1 2, 3 273
          NR_047588.1 4, 5 578
          NR_047589.1 3, 4 342
          NR_047590.1 4, 5 515
          NR_047591.1 4, 5 401
          NR_047592.1 4, 5 481
          XM_006718779.3 XP_006718842.1 6, 7 838
          XM_006718780.3 XP_006718843.1 5, 6 735
          XM_006718781.3 XP_006718844.1 6, 7 792
          XM_006718783.3 XP_006718846.1 4, 5 601
          XM_017017322.1 XP_016872811.1 5, 6 625
          XM_017017323.1 XP_016872812.1 4, 5 551
          XM_017017324.1 XP_016872813.1 4, 5 553
          GenBank mRNA AK091324.1 - 1, 2 3836
          AL833215.1 - 1, 2 240
          BC132716.1 - 4, 5 496 40146738
          BC132718.1 - 4, 5 496 40146740
          BC144270.1 - 5, 6 599 9052790
          BC144276.1 - 4, 5 449
          BC144277.1 - 4, 5 530 9052797
          BE513148.1 - 4, 5 469
          BE974033.1 - 2, 3 118
          BI192743.1 - 4, 5 382
          BI547955.1 - 4, 5 287
          BI856825.1 - 4, 5 403
          BM042315.1 - 5, 6 489
          BM928068.1 - 569
          BQ223298.1 - 6, 7 586
          BQ225039.1 - 3, 4 259
          BQ420663.1 - 4, 5 504
          BQ653243.1 - 4, 5 504
          BQ681518.1 - 5, 6 487
          BU160644.1 - 4, 5 334
          BU598207.1 - 4, 5 431
          BU855096.1 - 4, 5 425
          DA899308.1 - 4, 5 497

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0