Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM2D3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307960.1 | NP_001294889.1 | 2 | 202 | ||
NM_001308026.1 | NP_001294955.1 | 3 | 280 | |||
NM_025141.3 | NP_079417.2 | 2 | 202 | |||
NM_078474.2 | NP_510883.2 | 3 | 280 | |||
GenBank mRNA | AF353992.1 | - | 2 | 177 | ||
AK026257.1 | - | 2 | 192 | |||
AK094955.1 | - | 1 | 612 | |||
AK296553.1 | - | 3 | 271 | |||
AK297048.1 | - | 2 | 187 | |||
AK315584.1 | - | 3 | 256 | |||
BC006150.1 | - | 3 | 252 | 3051281 | ||
BC008873.2 | - | 2 | 175 | 4137334 | ||
BU595412.1 | - | 3 | 275 |