Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC727751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK291684.1 | - | 5 | 846 |
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026448.1 | - | 5 | 730 |
Gene Symbol: LOC101929479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022812.1 | XP_016878301.1 | 5 | 917 |