Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other VEZT SIRNA Assays ›
Gene Symbol:
VEZT
Gene Name:
vezatin, adherens junctions transmembrane protein
Gene Aliases:
VEZATIN
Chromosome Location:
Chr.12: 95217746 - 95302799 on Build GRCh38
UniGene ID:
Hs.24135
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31031
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: VEZT
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017599.3
NP_060069.3
10, 11
1736
NR_038241.1
11, 12
1809
NR_038242.1
10, 11
1744
XM_006719477.3
XP_006719540.1
10, 11
1748
XM_006719478.2
XP_006719541.1
9, 10
1642
XM_006719479.2
XP_006719542.1
11, 12
1819
XM_006719480.3
XP_006719543.1
11, 12
1796
XM_006719481.2
XP_006719544.1
10, 11
1666
XM_006719482.3
XP_006719545.1
10, 11
2047
XM_006719483.3
XP_006719546.1
10, 11
2064
XM_006719484.3
XP_006719547.1
9, 10
1974
XM_006719485.2
XP_006719548.1
8, 9
1416
XM_006719486.3
XP_006719549.1
8, 9
1404
XM_006719490.3
XP_006719553.1
11, 12
1807
XM_011538543.1
XP_011536845.1
11, 12
1741
XM_011538544.1
XP_011536846.1
11, 12
1729
XM_011538545.2
XP_011536847.1
10, 11
1709
XM_011538546.1
XP_011536848.1
12, 13
1812
XM_011538547.1
XP_011536849.1
12, 13
1856
XM_011538548.2
XP_011536850.1
9, 10
1502
XM_011538549.1
XP_011536851.1
11, 12
1741
XM_011538550.2
XP_011536852.1
11, 12
1741
XM_011538551.2
XP_011536853.1
11, 12
1729
XM_011538552.1
XP_011536854.1
11, 12
1815
XM_011538553.2
XP_011536855.1
11, 12
1745
XM_017019586.1
XP_016875075.1
10, 11
1697
XM_017019587.1
XP_016875076.1
9, 10
1630
XM_017019588.1
XP_016875077.1
12, 13
1800
XM_017019589.1
XP_016875078.1
11, 12
1784
XM_017019590.1
XP_016875079.1
12, 13
1844
XM_017019591.1
XP_016875080.1
10, 11
2035
XM_017019592.1
XP_016875081.1
9, 10
1489
XM_017019593.1
XP_016875082.1
12, 13
1864
XM_017019594.1
XP_016875083.1
11, 12
1726
XM_017019595.1
XP_016875084.1
10, 11
1748
XM_017019596.1
XP_016875085.1
10, 11
1748
XM_017019597.1
XP_016875086.1
10, 11
1736
XM_017019598.1
XP_016875087.1
10, 11
1736
XM_017019599.1
XP_016875088.1
10, 11
1697
XM_017019600.1
XP_016875089.1
10, 11
2052
XM_017019601.1
XP_016875090.1
11, 12
1803
XM_017019602.1
XP_016875091.1
12, 13
1800
XM_017019603.1
XP_016875092.1
10, 11
2064
XM_017019604.1
XP_016875093.1
8, 9
1416
XM_017019605.1
XP_016875094.1
8, 9
1404
GenBank mRNA
AF216644.1
-
10, 11
1663
AF225417.1
-
10, 11
1707
AF277625.1
-
6, 7
1175
AK001338.1
-
8, 9
1132
AK225303.1
-
11, 12
1749
AK225356.1
-
11, 12
1758
AK307649.1
-
11, 12
1815
AL133113.1
-
3, 4
462
AL137497.1
-
11, 12
1731
BC028597.1
-
13, 14
2065
BC064939.1
-
11, 12
1726
6159768
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
single organismal cell-cell adhesion
myosin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
git-commit: a47d3bef29014c2544d0032e3b149846d456d521
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0