Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HP1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016287.3 | NP_057371.2 | 6 | 678 | ||
XM_005245875.4 | XP_005245932.1 | 6 | 1253 | |||
XM_005245876.3 | XP_005245933.1 | 6 | 786 | |||
XM_005245877.4 | XP_005245934.1 | 6 | 1114 | |||
XM_005245878.4 | XP_005245935.1 | 6 | 1050 | |||
XM_005245879.4 | XP_005245936.1 | 5 | 577 | |||
XM_011541532.1 | XP_011539834.1 | 6 | 770 | |||
XM_011541533.1 | XP_011539835.1 | 6 | 967 | |||
XM_011541534.2 | XP_011539836.1 | 5 | 587 | |||
XM_017001393.1 | XP_016856882.1 | 6 | 733 | |||
XM_017001394.1 | XP_016856883.1 | 6 | 1098 | |||
GenBank mRNA | AF113534.1 | - | 5 | 608 | ||
AK023129.1 | - | 6 | 678 | |||
AK291252.1 | - | 6 | 656 | |||
AK304065.1 | - | 5 | 554 | |||
AL833978.1 | - | 2 | 494 | |||
BC045660.1 | - | 2 | 1310 | 5271429 | ||
BC046170.1 | - | 6 | 713 | |||
CK000759.1 | - | 3 | 340 |