Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NSMF SIRNA Assays ›
          Gene Symbol:
          NSMF
          Gene Name:
          NMDA receptor synaptonuclear signaling and neuronal migration factor
          Gene Aliases:
          HH9, NELF
          Chromosome Location:
          Chr.9: 137447570 - 137459357 on Build GRCh38
          UniGene ID:
          Hs.455336
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s24933

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NSMF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130969.1 NP_001124441.1 9, 10 1276
          NM_001130970.1 NP_001124442.1 8, 9 1207
          NM_001130971.1 NP_001124443.1 7, 8 1201
          NM_001178064.1 NP_001171535.1 8, 9 1186
          NM_015537.4 NP_056352.3 8, 9 1270
          XM_005266061.4 XP_005266118.1 7, 8 1203
          XM_005266062.4 XP_005266119.1 6, 7 1134
          XM_011518496.2 XP_011516798.1 7, 8 1140
          XM_011518497.2 XP_011516799.1 4, 5 390
          XM_017014597.1 XP_016870086.1 3, 4 659
          GenBank mRNA AJ420418.1 - 1, 2 51
          AK027474.1 - 6, 7 541
          AK074602.1 - 8, 9 1238
          AK292030.1 - 4, 5 342
          AK294320.1 - 4, 5 319
          AK303703.1 - 6, 7 398
          AK303976.1 - 4, 5 437
          AK316046.1 - 6, 7 592
          AL117660.2 - 4, 5 298
          AL389944.1 - 4, 5 324
          AY255128.1 - 9, 10 1276
          AY255129.1 - 8, 9 1270
          AY255130.1 - 8, 9 1207
          AY255131.1 - 8, 9 1186
          BC016862.2 - 4, 5 317
          BC072412.1 - 8, 9 1255 6165009
          BC110498.2 - 7, 8 1068 40007676

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein dephosphorylation
          regulation of neuron apoptotic process
          regulation of neuronal synaptic plasticity
          regulation of dendrite morphogenesis
          cellular response to amino acid stimulus
          cellular response to electrical stimulus
          cellular response to gonadotropin stimulus
          positive regulation of neuron migration
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0