Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047857.3 | NP_001041322.2 | 2, 3 | 568 | ||
XM_006234405.2 | XP_006234467.1 | 2, 3 | 495 |