Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287436.1 | NP_001274365.1 | 11 | 2283 | ||
NM_015198.3 | NP_056013.2 | 10 | 2112 | |||
XM_005271750.1 | XP_005271807.1 | 12 | 2319 | |||
XM_005271751.1 | XP_005271808.1 | 11 | 2274 | |||
XM_005271756.1 | XP_005271813.1 | 10 | 2103 | |||
XM_011515234.1 | XP_011513536.1 | 13 | 2394 | |||
XM_011515235.1 | XP_011513537.1 | 12 | 2349 | |||
XM_011515236.1 | XP_011513538.1 | 13 | 2394 | |||
XM_011515237.1 | XP_011513539.1 | 12 | 2223 | |||
XM_011515238.1 | XP_011513540.1 | 13 | 2394 | |||
XM_011515239.1 | XP_011513541.1 | 12 | 2223 | |||
XM_011515240.1 | XP_011513542.1 | 11 | 2178 | |||
XM_011515241.1 | XP_011513543.1 | 7 | 1398 | |||
XM_017011898.1 | XP_016867387.1 | 11 | 2148 | |||
XM_017011899.1 | XP_016867388.1 | 11 | 2274 | |||
XM_017011900.1 | XP_016867389.1 | 10 | 2103 | |||
GenBank mRNA | AB014533.1 | - | 10 | 2093 | ||
AK293883.1 | - | 3 | 649 | |||
AL713786.1 | - | 2 | 5567 | |||
BC029275.1 | - | 5 | 983 | |||
BC045771.1 | - | 9 | 1582 | |||
BC136441.1 | - | 11 | 2198 | 9020430 | ||
BC144099.1 | - | 10 | 2027 | 9052619 | ||
BC150263.1 | - | 10 | 2093 | 9007207 | ||
CR936689.1 | - | 9 | 1603 |