Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127453.1 | NP_001120925.1 | 3, 4 | 496 | ||
NM_001127454.1 | NP_001120926.1 | 2, 3 | 511 | |||
NM_004403.2 | NP_004394.1 | 3, 4 | 741 | |||
XM_017011802.1 | XP_016867291.1 | 2, 3 | 267 | |||
GenBank mRNA | AF007790.2 | - | 2, 3 | 396 | ||
AF073308.1 | - | 3, 4 | 465 | |||
AK094714.1 | - | 5, 6 | 1069 | |||
AK290775.1 | - | 3, 4 | 472 | |||
AK314402.1 | - | 3, 4 | 481 | |||
BC099911.1 | - | 3, 4 | 457 | 5287362 | ||
BC125065.1 | - | 3, 4 | 463 | 40117291 | ||
BC125066.1 | - | 2, 3 | 233 | |||
DA820184.1 | - | 3, 4 | 496 |