Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF415 SIRNA Assays ›
Gene Symbol:
ZNF415
Gene Name:
zinc finger protein 415
Gene Aliases:
ZfLp
Chromosome Location:
Chr.19: 53107879 - 53133077 on Build GRCh38
UniGene ID:
Hs.147765
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s230102
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF415
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136038.2
NP_001129510.2
4
1313
NM_001164309.1
NP_001157781.1
4
1186
NM_018355.3
NP_060825.2
4
1081
NR_028343.1
7
1442
XM_006723267.1
XP_006723330.1
6
1399
XM_006723268.1
XP_006723331.1
4
1147
XM_006723272.2
XP_006723335.1
5
1179
XM_006723273.2
XP_006723336.1
5
1263
XM_006723274.2
XP_006723337.1
5
1340
XM_006723277.2
XP_006723340.1
5
1274
XM_006723279.1
XP_006723342.1
5
1433
XM_006723280.2
XP_006723343.1
2
889
XM_006723281.2
XP_006723344.1
3
915
XM_006723282.1
XP_006723345.1
3
1224
XM_011527100.1
XP_011525402.1
5
1423
XM_011527101.2
XP_011525403.1
5
1398
XM_011527103.1
XP_011525405.1
5
1218
XM_011527104.1
XP_011525406.1
5
1512
XM_017026952.1
XP_016882441.1
5
1226
XM_017026953.1
XP_016882442.1
6
1546
XM_017026954.1
XP_016882443.1
6
1423
XM_017026955.1
XP_016882444.1
6
1374
XM_017026956.1
XP_016882445.1
7
1524
XM_017026957.1
XP_016882446.1
6
1262
XM_017026958.1
XP_016882447.1
7
1578
XM_017026959.1
XP_016882448.1
6
1449
XM_017026960.1
XP_016882449.1
5
1164
XM_017026961.1
XP_016882450.1
6
1610
XM_017026962.1
XP_016882451.1
6
1455
XM_017026963.1
XP_016882452.1
5
1351
XM_017026964.1
XP_016882453.1
5
1301
XM_017026965.1
XP_016882454.1
4
1315
XM_017026966.1
XP_016882455.1
5
1443
XM_017026967.1
XP_016882456.1
4
1241
XM_017026968.1
XP_016882457.1
6
1539
XM_017026969.1
XP_016882458.1
2
960
XM_017026970.1
XP_016882459.1
6
1416
XM_017026971.1
XP_016882460.1
6
1635
XM_017026972.1
XP_016882461.1
4
1217
XM_017026973.1
XP_016882462.1
5
1409
XM_017026974.1
XP_016882463.1
6
1488
GenBank mRNA
AK002053.1
-
4
1053
AK095504.1
-
7
1414
AY283600.1
-
4
1053
BC063880.1
-
5
1264
30388597
BC069668.1
-
5
1263
7262632
DQ925695.1
-
3
915
DQ925696.1
-
5
1252
DQ925697.1
-
6
1262
DQ925698.1
-
5
1364
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c48b94556-6l4lq:80/100.66.134.75:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0