Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRAK1 SIRNA Assays ›
          Gene Symbol:
          TRAK1
          Gene Name:
          trafficking kinesin protein 1
          Gene Aliases:
          MILT1, OIP106
          Chromosome Location:
          Chr.3: 42013802 - 42225890 on Build GRCh38
          UniGene ID:
          Hs.535711
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s22642

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRAK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042646.2 NP_001036111.1 8, 9 1123
          NM_001265608.1 NP_001252537.1 8, 9 1123
          NM_001265609.1 NP_001252538.1 7, 8 986
          NM_001265610.1 NP_001252539.1 7, 8 986
          NM_014965.4 NP_055780.2 7, 8 926
          XM_005264956.2 XP_005265013.1 8, 9 931
          XM_005264958.2 XP_005265015.1 7, 8 986
          XM_005264960.3 XP_005265017.1 8, 9 931
          XM_005264962.3 XP_005265019.1 8, 9 931
          XM_005264963.3 XP_005265020.1 8, 9 931
          XM_006713029.2 XP_006713092.1 8, 9 931
          XM_006713030.2 XP_006713093.1 8, 9 931
          XM_006713031.3 XP_006713094.1 8, 9 931
          XM_011533489.2 XP_011531791.1 8, 9 931
          XM_017005906.1 XP_016861395.1 10, 11 1544
          XM_017005907.1 XP_016861396.1 7, 8 908
          XM_017005908.1 XP_016861397.1 7, 8 986
          XM_017005909.1 XP_016861398.1 9, 10 1429
          XM_017005910.1 XP_016861399.1 7, 8 986
          XM_017005911.1 XP_016861400.1 9, 10 1429
          XM_017005912.1 XP_016861401.1 8, 9 931
          XM_017005913.1 XP_016861402.1 8, 9 931
          GenBank mRNA AB028965.1 - 8, 9 1123
          AK000754.1 - 7, 8 902
          AK294221.1 - 7, 8 952
          AK295848.1 - 8, 9 1030
          AK316260.1 - 8, 9 1020
          AL713787.1 - 7, 8 890
          BC015922.1 - 7, 8 913 4562916
          BX647199.1 - 7, 8 926

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          protein O-linked glycosylation
          protein targeting
          endosome to lysosome transport
          protein binding
          GABA receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0