Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RUFY3 SIRNA Assays ›
Gene Symbol:
RUFY3
Gene Name:
RUN and FYVE domain containing 3
Gene Aliases:
RIPX, SINGAR1, ZFYVE30
Chromosome Location:
Chr.4: 70704204 - 70808622 on Build GRCh38
UniGene ID:
Hs.740904
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22630
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RUFY3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001037442.3
NP_001032519.1
7, 8
1550
NM_001130709.1
NP_001124181.1
7, 8
998
NM_001291993.1
NP_001278922.1
8, 9
942
NM_001291994.1
NP_001278923.1
8, 9
1604
NM_014961.4
NP_055776.1
7, 8
1550
XM_005265656.2
XP_005265713.1
8, 9
1588
XM_005265658.1
XP_005265715.1
7, 8
864
XM_011531750.2
XP_011530052.1
8, 9
1186
XM_011531751.2
XP_011530053.1
7, 8
1199
XM_011531752.2
XP_011530054.1
7, 8
1199
XM_011531753.2
XP_011530055.1
7, 8
1199
XM_011531754.2
XP_011530056.1
7, 8
1199
XM_011531755.2
XP_011530057.1
7, 8
1164
XM_011531756.2
XP_011530058.1
7, 8
1164
XM_017007890.1
XP_016863379.1
7, 8
1199
XM_017007891.1
XP_016863380.1
8, 9
896
XM_017007892.1
XP_016863381.1
7, 8
864
GenBank mRNA
AB020678.1
-
7, 8
1356
AF112221.1
-
7, 8
780
AK000911.1
-
7, 8
804
AK294421.1
-
7, 8
735
AK296501.1
-
8, 9
791
AK302637.1
-
8, 9
818
BC051716.1
-
7, 8
1418
5265230
BC053837.1
-
7, 8
1380
BC071988.1
-
7, 8
1353
BC098433.1
-
7, 8
1380
DB482497.1
-
1, 2
16
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
actin filament organization
positive regulation of cell migration
positive regulation of axon extension
negative regulation of axonogenesis
positive regulation of intracellular protein transport
regulation of establishment of cell polarity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c48b94556-6l4lq:80/100.66.134.75:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0