Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF415 SIRNA Assays ›
Gene Symbol:
ZNF415
Gene Name:
zinc finger protein 415
Gene Aliases:
ZfLp
Chromosome Location:
Chr.19: 53107879 - 53133077 on Build GRCh38
UniGene ID:
Hs.147765
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s224389
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF415
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136038.2
NP_001129510.2
4
932
NM_001164309.1
NP_001157781.1
4
805
NM_018355.3
NP_060825.2
4
700
NR_028343.1
7
1061
XM_006723267.1
XP_006723330.1
6
1018
XM_006723268.1
XP_006723331.1
4
766
XM_006723272.2
XP_006723335.1
5
798
XM_006723273.2
XP_006723336.1
5
882
XM_006723274.2
XP_006723337.1
5
959
XM_006723277.2
XP_006723340.1
5
893
XM_006723279.1
XP_006723342.1
5
1052
XM_006723280.2
XP_006723343.1
2
508
XM_006723281.2
XP_006723344.1
3
534
XM_006723282.1
XP_006723345.1
3
843
XM_011527100.1
XP_011525402.1
5
1042
XM_011527101.2
XP_011525403.1
5
1017
XM_011527103.1
XP_011525405.1
5
837
XM_011527104.1
XP_011525406.1
5
1131
XM_017026952.1
XP_016882441.1
5
845
XM_017026953.1
XP_016882442.1
6
1165
XM_017026954.1
XP_016882443.1
6
1042
XM_017026955.1
XP_016882444.1
6
993
XM_017026956.1
XP_016882445.1
7
1143
XM_017026957.1
XP_016882446.1
6
881
XM_017026958.1
XP_016882447.1
7
1197
XM_017026959.1
XP_016882448.1
6
1068
XM_017026960.1
XP_016882449.1
5
783
XM_017026961.1
XP_016882450.1
6
1229
XM_017026962.1
XP_016882451.1
6
1074
XM_017026963.1
XP_016882452.1
5
970
XM_017026964.1
XP_016882453.1
5
920
XM_017026965.1
XP_016882454.1
4
934
XM_017026966.1
XP_016882455.1
5
1062
XM_017026967.1
XP_016882456.1
4
860
XM_017026968.1
XP_016882457.1
6
1158
XM_017026969.1
XP_016882458.1
2
579
XM_017026970.1
XP_016882459.1
6
1035
XM_017026971.1
XP_016882460.1
6
1254
XM_017026972.1
XP_016882461.1
4
836
XM_017026973.1
XP_016882462.1
5
1028
XM_017026974.1
XP_016882463.1
6
1107
GenBank mRNA
AK002053.1
-
4
672
AL534796.3
-
5
882
AY283600.1
-
4
672
BC063880.1
-
5
883
30388597
BC069668.1
-
5
882
7262632
DQ925695.1
-
3
534
DQ925696.1
-
5
871
DQ925697.1
-
6
881
DQ925698.1
-
5
983
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c48b94556-6l4lq:80/100.66.134.75:80.
git-commit: 256cec658627a60c8e1196038bf5905a13cd45ba
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0