Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GATA4 SIRNA Assays ›
          Gene Symbol:
          Gata4
          Gene Name:
          GATA binding protein 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 46386703 - 46432914 on Build Rnor_6.0
          UniGene ID:
          Rn.26251
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s222027

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gata4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_144730.1 NP_653331.1 3 1260
          XM_006252189.2 XP_006252251.1 2 840
          GenBank mRNA L22761.1 - 3 1260

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          gastrulation with mouth forming second
          endoderm formation
          heart looping
          heart morphogenesis
          atrioventricular valve morphogenesis
          atrioventricular valve formation
          endocardial cushion development
          cardiac right ventricle morphogenesis
          ventricular cardiac muscle tissue development
          cardiac septum development
          ventricular septum development
          atrial septum primum morphogenesis
          atrial septum secundum morphogenesis
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cell-cell signaling
          spermatogenesis
          heart development
          male gonad development
          response to mechanical stimulus
          regulation of gene expression
          negative regulation of autophagy
          positive regulation of vascular endothelial growth factor production
          cardiac muscle hypertrophy in response to stress
          signal transduction involved in regulation of gene expression
          BMP signaling pathway
          positive regulation of BMP signaling pathway
          response to retinoic acid
          response to vitamin A
          embryonic heart tube development
          embryonic heart tube anterior/posterior pattern specification
          tube morphogenesis
          atrioventricular canal development
          response to drug
          response to estrogen
          positive regulation of angiogenesis
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          embryonic digestive tract morphogenesis
          embryonic morphogenesis
          embryonic foregut morphogenesis
          cardiac muscle tissue development
          positive regulation of cardioblast differentiation
          cardiac muscle cell differentiation
          Sertoli cell differentiation
          regulation of cardiac muscle cell proliferation
          positive regulation of cardiac muscle cell proliferation
          canonical Wnt signaling pathway
          transdifferentiation
          SMAD protein signal transduction
          atrial septum morphogenesis
          lung lobe formation
          intestinal epithelial cell differentiation
          cell growth involved in cardiac muscle cell development
          cellular response to glucose stimulus
          cellular response to gonadotropin stimulus
          cellular response to follicle-stimulating hormone stimulus
          epithelial cell fate commitment
          seminiferous tubule development
          regulation of cardiac muscle cell contraction
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II transcription factor binding
          enhancer sequence-specific DNA binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
          transcription coactivator activity
          transcription factor binding
          zinc ion binding
          protein kinase binding
          activating transcription factor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          NFAT protein binding
          co-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0