Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other B2M SIRNA Assays ›
          Gene Symbol:
          B2m
          Gene Name:
          beta-2 microglobulin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 114087287 - 114093311 on Build Rnor_6.0
          UniGene ID:
          Rn.1868
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s217816

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: B2m
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012512.2 NP_036644.1 2 258
          GenBank mRNA FM032790.1 - 2 258
          FQ210051.1 - 2 271
          FQ210252.1 - 2 272
          FQ210906.1 - 2 271
          FQ211296.1 - 2 271
          FQ211419.1 - 2 270
          FQ212410.1 - 2 272
          FQ217926.1 - 2 269
          FQ218141.1 - 2 271
          FQ218251.1 - 2 271
          FQ218298.1 - 2 270
          FQ219455.1 - 820
          FQ220256.1 - 2 267
          FQ221268.1 - 2 271
          FQ221287.1 - 2 267
          FQ221985.1 - 2 270
          FQ222352.1 - 2 268
          FQ225010.1 - 2 271
          FQ225051.1 - 2 275
          FQ225087.1 - 2 270
          FQ225088.1 - 2 272
          FQ225121.1 - 2 270
          FQ225141.1 - 2 270
          FQ225219.1 - 2 269
          FQ225292.1 - 2 271
          FQ225550.1 - 2 270
          FQ225647.1 - 2 270
          FQ225686.1 - 2 273
          FQ225690.1 - 2 412
          FQ225847.1 - 2 270
          FQ225912.1 - 2 270
          FQ226004.1 - 2 271
          FQ226258.1 - 2 273
          FQ226642.1 - 2 272
          FQ226723.1 - 2 270
          FQ227149.1 - 2 270
          FQ227291.1 - 2 276
          FQ227327.1 - 2 271
          FQ227454.1 - 2 271
          FQ227468.1 - 2 269
          FQ227656.1 - 2 271
          FQ227700.1 - 2 271
          FQ227950.1 - 2 269
          FQ227964.1 - 2 270
          FQ227995.1 - 2 266
          FQ228065.1 - 2 272
          FQ228188.1 - 2 271
          FQ228427.1 - 2 272
          FQ229482.1 - 2 263
          FQ229718.1 - 2 270
          FQ230202.1 - 2 271
          FQ230402.1 - 2 271
          FQ230977.1 - 2 270
          FQ231135.1 - 2 271
          FQ231182.1 - 2 271
          FQ231367.1 - 2 270
          FQ231378.1 - 2 270
          FQ231524.1 - 2 270
          FQ231602.1 - 2 271
          FQ231718.1 - 2 270
          FQ231856.1 - 2 270
          FQ231894.1 - 2 270
          FQ232233.1 - 2 271
          FQ232294.1 - 2 271
          FQ232460.1 - 2 270
          FQ232758.1 - 2 270
          FQ232818.1 - 2 270
          FQ232819.1 - 2 270
          FQ232937.1 - 2 270
          FQ232943.1 - 2 272
          FQ232957.1 - 2 268
          FQ233220.1 - 2 271
          FQ233240.1 - 2 271
          FQ233290.1 - 2 270
          FQ233447.1 - 2 271
          FQ233505.1 - 2 270
          FQ233544.1 - 2 270
          FQ233567.1 - 2 275
          FQ233757.1 - 2 270
          FQ233797.1 - 2 269
          FQ233849.1 - 2 270
          FQ234348.1 - 2 271
          FQ234349.1 - 2 270
          FQ234524.1 - 2 268
          FQ234581.1 - 2 268
          FQ234840.1 - 2 270
          FQ234885.1 - 2 270
          FQ235105.1 - 2 270
          FQ235149.1 - 2 268
          FQ235229.1 - 2 270
          X16956.1 - 1 47
          Y00441.1 - 2 245
          Y08531.1 - 2 250

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_004048, DQ839493, AB021288, S54761, S82297, and others.
          This siRNA also targets the following transcripts in Mouse: NM_009735, BC085164, AK020632, X01838, AK019389, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          positive regulation of T cell mediated cytotoxicity
          response to molecule of bacterial origin
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
          T cell differentiation in thymus
          protein refolding
          response to drug
          response to cadmium ion
          iron ion homeostasis
          molecular_function
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-89bfcd4dd-zgn5p:80/100.66.134.75:80.
          git-commit: f1ed3cb73da2dc18796bd96aca0ec275a318700b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0