Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgfr1op2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026218.2 | NP_080494.1 | 3 | 458 | ||
GenBank mRNA | AK004662.1 | - | 3 | 442 | ||
AK005320.1 | - | 3 | 435 | |||
AK008983.1 | - | 3 | 442 | |||
AK050102.1 | - | 3 | 458 | |||
AK151318.1 | - | 3 | 451 | |||
AK152617.1 | - | 3 | 451 | |||
AK153137.1 | - | 3 | 451 | |||
AK159805.1 | - | 3 | 407 | |||
AK169380.1 | - | 3 | 467 | |||
AK169383.1 | - | 3 | 435 | |||
BC028876.1 | - | 3 | 421 | 4504364 |