Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006369.4 | NP_006360.3 | 6, 7 | 1974 | ||
XM_017000074.1 | XP_016855563.1 | 6, 7 | 2000 | |||
GenBank mRNA | AF370430.1 | - | 6, 7 | 1874 | ||
AK125827.1 | - | 5, 6 | 3251 | |||
AK223420.1 | - | 3, 4 | 1336 | |||
AK291283.1 | - | 6, 7 | 1957 | |||
AK296436.1 | - | 6, 7 | 1793 | |||
AK297596.1 | - | 6, 7 | 1988 | |||
BC004953.1 | - | 3, 4 | 1210 | |||
BC101522.1 | - | 6, 7 | 1964 | 8069028 | ||
BC101524.1 | - | 6, 7 | 1964 | 8069030 | ||
BX537366.1 | - | 6, 7 | 1884 | |||
X86018.1 | - | 3, 4 | 1186 |