Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007938.2 | NP_031964.2 | 5 | 1609 | ||
GenBank mRNA | BC141090.1 | - | 5 | 1125 | 9055702 | |
BC145362.1 | - | 6 | 1245 | |||
U58332.1 | - | 5 | 1325 |