Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr137 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109562.2 | NP_001103032.1 | 1 | 500 | ||
GenBank mRNA | BC167037.1 | - | 1 | 500 | 7462064 |