Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uchl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110165.1 | NP_001103635.1 | 7, 8 | 630 | ||
GenBank mRNA | AB043959.1 | - | 7, 8 | 555 |