Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Purb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017503.1 | NP_001017503.1 | 1 | 905 | ||
GenBank mRNA | AB182574.1 | - | 1 | 905 |