Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012631.1 | NP_001012649.1 | 7, 8 | 489 | ||
NM_001012632.1 | NP_001012650.1 | 6, 7 | 288 | |||
NM_001012633.1 | NP_001012651.1 | 6, 7 | 422 | |||
NM_001012634.1 | NP_001012652.1 | 5, 6 | 229 | |||
NM_001012635.1 | NP_001012653.1 | 5, 6 | 224 | |||
NM_001012636.1 | NP_001012654.1 | 4, 5 | 212 | |||
NM_001012718.1 | NP_001012736.1 | 6, 7 | 425 | |||
NM_001308078.1 | NP_001295007.1 | 5, 6 | 423 | |||
NM_004221.4 | NP_004212.4 | 6, 7 | 422 | |||
GenBank mRNA | AK296143.1 | - | 5, 6 | 234 | ||
AY495331.1 | - | 6, 7 | 412 | |||
AY495332.1 | - | 6, 7 | 412 | |||
AY495333.1 | - | 5, 6 | 370 | |||
BC009401.2 | - | 5, 6 | 560 | 4300407 | ||
BC018782.1 | - | 6, 7 | 269 | 4873608 | ||
BC105602.1 | - | 6, 7 | 290 | 6496101 | ||
BF568456.1 | - | 5, 6 | 561 | |||
BG748458.1 | - | 6, 7 | 368 | |||
BM759359.1 | - | 3, 4 | 163 | |||
BM822755.1 | - | 7, 8 | 397 | |||
JN546102.1 | - | 4, 5 | 151 | |||
M59807.1 | - | 5, 6 | 408 |