Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cers5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108993.1 | NP_001102463.1 | 5 | 658 | ||
XM_006257374.2 | XP_006257436.1 | 5 | 641 | |||
XM_006257375.2 | XP_006257437.1 | 3 | 500 | |||
GenBank mRNA | BC166794.1 | - | 5 | 540 | 7937721 |