Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sms | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033899.1 | NP_001029071.1 | 10, 11 | 1084 | ||
GenBank mRNA | BC101870.1 | - | 10, 11 | 1084 | 5621864 |