Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtpbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013225.2 | NP_001013243.2 | 6 | 872 | ||
XM_006244543.2 | XP_006244605.1 | 6 | 914 | |||
XM_006244544.2 | XP_006244606.1 | 6 | 704 | |||
XM_006244545.2 | XP_006244607.1 | 5 | 2788 | |||
XM_006244546.2 | XP_006244608.1 | 6 | 693 |