Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108754.1 | NP_001102224.1 | 7 | 851 | ||
GenBank mRNA | FQ215152.1 | - | 7 | 717 | ||
FQ226445.1 | - | 7 | 717 |