Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108333.1 | NP_001101803.1 | 3 | 708 | ||
XM_006249238.2 | XP_006249300.1 | 4 | 803 | |||
XM_006249239.2 | XP_006249301.1 | 4 | 555 | |||
XM_006249240.2 | XP_006249302.1 | 4 | 792 | |||
XM_008769150.1 | XP_008767372.1 | 4 | 799 | |||
GenBank mRNA | BC168697.1 | - | 1 | 107 |