Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggnbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004273.1 | NP_001004273.1 | 6 | 925 | ||
XM_006247047.2 | XP_006247109.1 | 8 | 1133 | |||
XM_006247048.2 | XP_006247110.1 | 8 | 1132 | |||
XM_006247049.2 | XP_006247111.1 | 8 | 1130 | |||
XM_006247050.2 | XP_006247112.1 | 8 | 1129 | |||
XM_006247051.2 | XP_006247113.1 | 7 | 1005 | |||
XM_006247052.2 | XP_006247114.1 | 7 | 1005 | |||
XM_006247053.2 | XP_006247115.1 | 7 | 1002 | |||
XM_006247054.2 | XP_006247116.1 | 7 | 1001 | |||
XM_008768070.1 | XP_008766292.1 | 8 | 1107 | |||
XM_008768071.1 | XP_008766293.1 | 8 | 1077 | |||
GenBank mRNA | AY633742.1 | - | 6 | 925 | ||
FQ220883.1 | - | 4 | 297 | |||
FQ232770.1 | - | 3 | 251 | |||
FQ234201.1 | - | 4 | 392 | |||
FQ234847.1 | - | 3 | 251 |