Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114633.1 | NP_001108105.1 | 14 | 1437 | ||
GenBank mRNA | AK124489.1 | - | 6 | 1748 | ||
AK299419.1 | - | 14 | 1403 | |||
BX648318.1 | - | 3 | 2415 | |||
CA488226.1 | - | 6 | 684 |
Gene Symbol: JMJD7-PLA2G4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198588.1 | NP_001185517.1 | 19 | 2069 | ||
NM_005090.3 | NP_005081.1 | 19 | 2069 | |||
GenBank mRNA | AB209150.1 | - | 19 | 5912 | ||
AF065215.1 | - | 19 | 2044 | |||
AF065216.1 | - | 20 | 5472 | |||
AF121908.1 | - | 19 | 2036 | |||
DQ523799.1 | - | 19 | 2037 | |||
DQ523800.1 | - | 19 | 2037 |