Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp26b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181087.2 | NP_851601.1 | 5, 6 | 1298 | ||
GenBank mRNA | AY245532.2 | - | 5, 6 | 1298 |