Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232299.2 | XP_006232361.1 | 2, 3 | 366 | ||
GenBank mRNA | BC166450.1 | - | 2, 3 | 306 | 8371068 |