Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107457.1 | NP_001100927.1 | 2 | 324 | ||
GenBank mRNA | BC166768.1 | - | 296 | 7933544 |