Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107255.1 | NP_001100725.1 | 12 | 1566 | ||
XM_006251795.2 | XP_006251857.1 | 12 | 1568 |