Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106923.1 | NP_001100393.1 | 9 | 1372 | ||
XM_008767226.1 | XP_008765448.1 | 10 | 1524 | |||
XM_008767227.1 | XP_008765449.1 | 9 | 2248 | |||
XM_008767228.1 | XP_008765450.1 | 9 | 2739 | |||
XM_008767229.1 | XP_008765451.1 | 9 | 2536 | |||
GenBank mRNA | BC168969.1 | - | 9 | 1377 | 9026830 |