Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other OSR1 SIRNA Assays ›
          Gene Symbol:
          Osr1
          Gene Name:
          odd-skipped related transciption factor 1
          Gene Aliases:
          Odd1
          Chromosome Location:
          Chr.6: 35314431 - 35321458 on Build Rnor_6.0
          UniGene ID:
          Rn.160611
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s151795

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Osr1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001106716.2 NP_001100186.1 1 335
          XM_006239886.2 XP_006239948.1 2 629
          GenBank mRNA BC159410.1 - 1 335 8366695

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          urogenital system development
          metanephros development
          ureteric bud development
          mesonephros development
          chondrocyte differentiation
          transcription, DNA-templated
          heart development
          gonad development
          positive regulation of gene expression
          cell differentiation
          positive regulation of bone mineralization
          negative regulation of epithelial cell differentiation
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          embryonic skeletal limb joint morphogenesis
          middle ear morphogenesis
          odontogenesis
          embryonic digit morphogenesis
          negative regulation of apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          intermediate mesoderm development
          pronephros development
          stem cell differentiation
          positive regulation of epithelial cell proliferation
          palate development
          embryonic skeletal joint morphogenesis
          cellular response to retinoic acid
          metanephric mesenchyme development
          cell proliferation involved in kidney development
          metanephric mesenchyme morphogenesis
          mesangial cell development
          metanephric mesenchymal cell differentiation
          posterior mesonephric tubule development
          specification of anterior mesonephric tubule identity
          specification of posterior mesonephric tubule identity
          mesonephric duct morphogenesis
          negative regulation of nephron tubule epithelial cell differentiation
          renal vesicle progenitor cell differentiation
          ureter urothelium development
          metanephric epithelium development
          metanephric smooth muscle tissue development
          metanephric nephron tubule development
          metanephric glomerulus vasculature development
          metanephric interstitial fibroblast development
          pattern specification involved in metanephros development
          embryonic skeletal joint development
          metanephric cap mesenchymal cell proliferation involved in metanephros development
          positive regulation of gastrulation
          nucleic acid binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7967b4b575-m9zbg:80/100.66.134.75:80.
          git-commit: a47d3bef29014c2544d0032e3b149846d456d521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0