Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF133 SIRNA Assays ›
Gene Symbol:
ZNF133
Gene Name:
zinc finger protein 133
Gene Aliases:
ZNF150, pHZ-13, pHZ-66
Chromosome Location:
Chr.20: 18288228 - 18316996 on Build GRCh38
UniGene ID:
Hs.472221
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s15167
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF133
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001083330.2
NP_001076799.1
4
1178
NM_001282995.1
NP_001269924.1
6
1414
NM_001282996.1
NP_001269925.1
6
1288
NM_001282997.1
NP_001269926.1
5
1339
NM_001282998.1
NP_001269927.1
4
1168
NM_001282999.1
NP_001269928.1
4
1181
NM_001283000.1
NP_001269929.1
5
1357
NM_001283001.1
NP_001269930.1
5
1344
NM_001283002.1
NP_001269931.1
5
1246
NM_001283003.1
NP_001269932.1
5
1319
NM_001283004.1
NP_001269933.1
5
1306
NM_001283005.1
NP_001269934.1
2
942
NM_001283006.1
NP_001269935.1
3
1020
NM_001283007.1
NP_001269936.1
3
1038
NM_001283008.1
NP_001269937.1
4
1208
NM_003434.5
NP_003425.2
7
1590
XM_005260819.1
XP_005260876.1
4
990
XM_005260820.4
XP_005260877.1
6
1247
XM_011529336.1
XP_011527638.1
7
1326
XM_011529337.1
XP_011527639.1
6
1198
XM_011529338.2
XP_011527640.1
6
1192
XM_011529339.1
XP_011527641.1
7
1326
XM_011529340.1
XP_011527642.1
2
801
XM_017028041.1
XP_016883530.1
5
9091
XM_017028042.1
XP_016883531.1
5
2365
XM_017028043.1
XP_016883532.1
5
9088
XM_017028044.1
XP_016883533.1
7
9438
XM_017028045.1
XP_016883534.1
5
9506
XM_017028046.1
XP_016883535.1
5
9104
XM_017028047.1
XP_016883536.1
5
2458
XM_017028048.1
XP_016883537.1
4
2287
XM_017028049.1
XP_016883538.1
7
9448
XM_017028050.1
XP_016883539.1
7
1346
XM_017028051.1
XP_016883540.1
8
1522
XM_017028052.1
XP_016883541.1
6
1186
XM_017028053.1
XP_016883542.1
4
1220
XM_017028054.1
XP_016883543.1
5
1100
XM_017028055.1
XP_016883544.1
8
1516
XM_017028056.1
XP_016883545.1
10
1735
XM_017028057.1
XP_016883546.1
7
1303
XM_017028058.1
XP_016883547.1
8
1520
GenBank mRNA
AK123005.1
-
2
2756
AK291389.1
-
5
1096
AK294263.1
-
3
769
AK295273.1
-
5
1018
AK295513.1
-
5
1082
AK313525.1
-
4
917
AK316386.1
-
2
696
BC001887.1
-
7
1396
3535533
BT007310.1
-
3
837
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.34.0-2025.08.32-1.0