Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpatch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024979.1 | NP_001020150.1 | 6, 7 | 546 | ||
XM_008761129.1 | XP_008759351.1 | 7, 8 | 1077 | |||
XM_008761130.1 | XP_008759352.1 | 7, 8 | 1127 | |||
XM_008761131.1 | XP_008759353.1 | 6, 7 | 1063 | |||
XM_008761132.1 | XP_008759354.1 | 7, 8 | 1241 | |||
GenBank mRNA | BC093381.1 | - | 6, 7 | 546 | 7388663 | |
FQ233538.1 | - | 3, 4 | 268 |