Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1305455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024969.2 | NP_001020140.2 | 2 | 625 | ||
GenBank mRNA | BC091263.1 | - | 2 | 625 | ||
FQ211718.1 | - | 2 | 671 |