Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009626.1 | NP_001009626.1 | 7 | 882 | ||
GenBank mRNA | BC088180.1 | - | 7 | 882 | 7364928 | |
FQ209412.1 | - | 7 | 921 | |||
FQ209491.1 | - | 7 | 908 | |||
FQ209740.1 | - | 7 | 918 | |||
FQ210182.1 | - | 7 | 909 | |||
FQ210311.1 | - | 7 | 908 | |||
FQ210756.1 | - | 7 | 924 | |||
FQ210813.1 | - | 7 | 908 | |||
FQ211328.1 | - | 7 | 881 | |||
FQ218068.1 | - | 2 | 195 | |||
FQ218335.1 | - | 3 | 415 | |||
FQ218445.1 | - | 7 | 922 | |||
FQ218454.1 | - | 7 | 909 | |||
FQ218597.1 | - | 7 | 904 | |||
FQ218599.1 | - | 7 | 908 | |||
FQ218637.1 | - | 7 | 908 | |||
FQ218785.1 | - | 7 | 925 | |||
FQ218859.1 | - | 7 | 881 | |||
FQ218865.1 | - | 7 | 908 | |||
FQ218961.1 | - | 7 | 908 | |||
FQ219138.1 | - | 7 | 973 | |||
FQ219328.1 | - | 7 | 908 | |||
FQ219380.1 | - | 7 | 909 | |||
FQ219438.1 | - | 7 | 907 | |||
FQ219481.1 | - | 7 | 920 | |||
FQ219537.1 | - | 7 | 905 | |||
FQ219580.1 | - | 7 | 922 | |||
FQ219601.1 | - | 7 | 921 | |||
FQ219674.1 | - | 7 | 908 | |||
X15551.1 | - | 7 | 737 |