Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134353.3 | NP_599180.1 | 9 | 1458 | ||
GenBank mRNA | AJ298278.1 | - | 9 | 1458 | ||
BC083176.1 | - | 9 | 1702 | 7097075 | ||
FQ225136.1 | - | 2 | 235 | |||
FQ225513.1 | - | 2 | 171 | |||
FQ225570.1 | - | 3 | 304 | |||
FQ225622.1 | - | 3 | 354 | |||
FQ226477.1 | - | 4 | 447 | |||
FQ226807.1 | - | 3 | 374 | |||
FQ227116.1 | - | 2 | 300 | |||
FQ227215.1 | - | 3 | 396 | |||
FQ227400.1 | - | 4 | 418 | |||
FQ227801.1 | - | 3 | 345 | |||
FQ227809.1 | - | 2 | 115 | |||
FQ228174.1 | - | 3 | 359 | |||
FQ230227.1 | - | 4 | 482 | |||
FQ230279.1 | - | 3 | 333 | |||
FQ231168.1 | - | 6 | 742 | |||
FQ231494.1 | - | 4 | 477 | |||
FQ231712.1 | - | 2 | 297 | |||
FQ231724.1 | - | 3 | 370 | |||
FQ232521.1 | - | 3 | 359 | |||
FQ232597.1 | - | 2 | 235 | |||
FQ233029.1 | - | 3 | 356 | |||
FQ233189.1 | - | 3 | 371 | |||
FQ233212.1 | - | 2 | 171 | |||
FQ233332.1 | - | 2 | 169 | |||
FQ233392.1 | - | 2 | 282 | |||
FQ233536.1 | - | 2 | 171 | |||
FQ233735.1 | - | 2 | 206 | |||
FQ233754.1 | - | 5 | 587 | |||
FQ234055.1 | - | 2 | 103 | |||
FQ234227.1 | - | 3 | 357 | |||
FQ234338.1 | - | 2 | 194 | |||
FQ234422.1 | - | 2 | 282 | |||
FQ234731.1 | - | 6 | 675 | |||
FQ235211.1 | - | 4 | 401 | |||
FQ235339.1 | - | 6 | 735 |