Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgst1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134349.3 | NP_599176.1 | 2 | 122 | ||
GenBank mRNA | BC063150.1 | - | 2 | 120 | 6922267 | |
BP494884.1 | - | 2 | 121 | |||
CB711033.1 | - | 2 | 138 | |||
FJ179404.1 | - | 1 | 51 | |||
FQ209427.1 | - | 2 | 143 | |||
FQ209464.1 | - | 2 | 135 | |||
FQ209465.1 | - | 2 | 142 | |||
FQ211049.1 | - | 2 | 142 | |||
FQ212841.1 | - | 2 | 143 | |||
FQ218096.1 | - | 2 | 125 | |||
FQ218830.1 | - | 2 | 150 | |||
FQ219010.1 | - | 2 | 144 | |||
FQ219410.1 | - | 2 | 116 | |||
FQ219464.1 | - | 2 | 148 | |||
FQ219581.1 | - | 2 | 144 | |||
FQ219850.1 | - | 2 | 134 | |||
J03752.1 | - | 2 | 106 |