Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CLASP2 SIRNA Assays ›
          Gene Symbol:
          Clasp2
          Gene Name:
          cytoplasmic linker associated protein 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 122003718 - 122185664 on Build Rnor_6.0
          UniGene ID:
          Rn.19943
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID s137697

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Clasp2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053722.2 NP_446174.1 3, 4 373
          XM_006243899.2 XP_006243961.1 9, 10 1035
          XM_006243901.2 XP_006243963.1 9, 10 1035
          XM_006243904.2 XP_006243966.1 9, 10 1035
          XM_006243905.2 XP_006243967.1 9, 10 1035
          XM_006243906.2 XP_006243968.1 9, 10 1035
          XM_006243910.2 XP_006243972.1 3, 4 1283
          XM_006243912.2 XP_006243974.1 3, 4 1280
          XM_006243914.2 XP_006243976.1 9, 10 1149
          XM_008766606.1 XP_008764828.1 9, 10 1035
          XM_008766607.1 XP_008764829.1 9, 10 1035
          XM_008766608.1 XP_008764830.1 9, 10 1035
          XM_008766609.1 XP_008764831.1 9, 10 1035
          XM_008766610.1 XP_008764832.1 9, 10 1035
          XM_008766611.1 XP_008764833.1 9, 10 1035
          XM_008766612.1 XP_008764834.1 9, 10 1035
          XM_008766613.1 XP_008764835.1 3, 4 1285
          XM_008766614.1 XP_008764836.1 3, 4 1552
          XM_008766615.1 XP_008764837.1 9, 10 1035
          GenBank mRNA AJ288060.1 - 3, 4 373

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001081960, NM_001286599, NM_001286603, NM_001286601, NM_029633, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule nucleation
          negative regulation of microtubule depolymerization
          mitotic nuclear division
          establishment or maintenance of cell polarity
          cell migration
          microtubule organizing center organization
          regulation of microtubule polymerization or depolymerization
          regulation of microtubule-based process
          microtubule anchoring
          cell division
          microtubule binding
          microtubule plus-end binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-854c5b95bd-957wg:80/100.66.134.75:80.
          git-commit: 47437712f32dbf53ebfc670c484059c2aca19dcb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0