Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031807.1 | NP_113995.1 | 2 | 1567 | ||
XM_006243499.2 | XP_006243561.1 | 2 | 1857 | |||
XM_008766457.1 | XP_008764679.1 | 2 | 1668 | |||
GenBank mRNA | AF063939.1 | - | 2 | 1567 | ||
BC087011.1 | - | 2 | 1567 | 7193411 |