Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031107.1 | NP_112369.1 | 13 | 1192 | ||
XM_006239083.2 | XP_006239145.1 | 12 | 1596 | |||
XM_006239084.2 | XP_006239146.1 | 12 | 1512 | |||
GenBank mRNA | M99169.1 | - | 13 | 1192 |